missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Standard For ICP, 10,000μg/mL in 5% HCl, SPEX CertiPrep™

Product Code. 10335892
Click to view available options
Quantity:
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Mg10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 7791-18-6, 7732-18-5
Packaging Triple-leached LDPE bottle
Quantity 500 mL
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Molecular Formula Cl2H12MgO6
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
IUPAC Name magnesium(2+) hexahydrate dichloride
Molecular Weight (g/mol) 203.30
Grade Assurance
Solvent or Matrix 5% HCl
Chemical Name or Material Magnesium Standard
Show More Show Less
compliance-icons
Product Identifier
  • 10,000 µg/mL Magnesium
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid,magnesium

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'