Learn More
Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 196105000
46.59 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
Specifications
Magnesium standard solution | |
7439-95-4, 7697-37-2, 7732-18-5 | |
500 mL | |
Mg | |
Liquid | |
997 to 1003 μg/mL of Mg (2+) in 0.5N HNO3 (20°C) | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
magnesium | |
24.30 | |
CHEBI:25107 | |
AAS |
1 mg/ml Mg in 0.5N HNO3 | |
Plastic bottle | |
1.74 | |
MFCD00085308 | |
16,198; 16,205; 16,356; 01,990; 04,422; 05,575; 06,507; 08,439; 10,345; 11,465; 14,281 | |
Colorless | |
[Mg] | |
Soluble in water | |
5462224 | |
24.31 |
Safety and Handling
- Magnesium atomic absorption standard solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only