missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate, 7-Hydrate, Crystal, Multi-Compendial, U.S.P., J.T. Baker™
603.25 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Physical Form | Crystals |
Quantity | 2.5 kg |
Formula Weight | 246.47 |
Grade | Multi-Compendial/USP |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.67kg |
Chemical Name or Material | Magnesium Sulfate, 7-Hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'