missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate, 97%, pure, anhydrous, Thermo Scientific Chemicals

Magnesium sulfate, 97%, MgO4S, CAS Number-7487-88-9, sulfuric acid magnesium salt 1:1, sal de sedlitz, magnesium sulfate, tomix ot, bitter salt, magnesium sulfate 1:1, sal angalis, magnesium sulphate, mgso4, magnesium sulfate anhydrous, 500g, CHEBI:32599, Undesignated, >700 deg.C

£18.39 - £1113.54

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
IUPAC Name magnesium;sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10625871
View Documents
Thermo Scientific Acros
413480050
5 g Glass Bottle
£18.39
5g
Please to purchase this item. Need a web account? Register with us today!
10731252
View accessory products View Documents Promotion Details
Thermo Scientific Acros
413485000
500 g Plastic bottle
£27.03
500g
Please to purchase this item. Need a web account? Register with us today!
10003812
View Documents Promotion Details
Thermo Scientific Acros
413480025
2.5 kg Plastic Bottle
£126.63
2.50kg
Please to purchase this item. Need a web account? Register with us today!
10107253
View Documents
Thermo Scientific Acros
413480250
25 kg Fibre drum
£1113.54
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
Specifications

Chemical Identifiers

7487-88-9
120.36
CSNNHWWHGAXBCP-UHFFFAOYSA-L
24083
magnesium;sulfate
MgO4S
MFCD00011110
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
CHEBI:32599
[Mg++].[O-]S([O-])(=O)=O

Specifications

1124.0°C
Granular Powder and/or Specks
MgO4S
MFCD00011110
15, 5755
Solubility in water: 26g/100mL (0°C).
[Mg++].[O-]S([O-])(=O)=O
120.36
CHEBI:32599
97%
>700°C
7487-88-9
97%
MgSO4
01,634; 04,425; 05,421
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
CSNNHWWHGAXBCP-UHFFFAOYSA-L
magnesium;sulfate
24083
120.37
Pure
Magnesium sulfate, Pure
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only