missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Extra Pure, SLR, Dried, Fisher Chemical™

Product Code. 10224680
Click to view available options
Quantity:
500 g
1 kg
2 kg
10 kg
Packaging:
Plastic keg
Plastic powder jar

Magnesium Sulfate, Extra Pure, SLR, Dried

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22189-08-8
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00149787
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1124°C
Color White
Physical Form Powder/Solid
Quantity 2 kg
IUPAC Name magnesium;sulfate
Formula Weight 120.36g/mol
Packaging Plastic powder jar
Vapor Pressure <0.1mmHg at 20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.