missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate, Extra Pure, SLR, Dried, Fisher Chemical™
29.39 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
2 kg
10 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Magnesium Sulfate, Extra Pure, SLR, Dried
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 1124°C |
Color | White |
Physical Form | Powder/Solid |
Quantity | 500 g |
IUPAC Name | magnesium;sulfate |
Formula Weight | 120.36g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | <0.1mmHg at 20°C |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'