missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium sulfate heptahydrate, 99+%, extra pure, Thermo Scientific Chemicals
£45.10 - £247.00
Chemical Identifiers
CAS | 10034-99-8 |
---|---|
Molecular Formula | H14MgO11S |
Molecular Weight (g/mol) | 246.47 |
MDL Number | MFCD00149785 |
InChI Key | WRUGWIBCXHJTDG-UHFFFAOYSA-L |
Synonym | magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate |
PubChem CID | 24843 |
ChEBI | CHEBI:31795 |
SMILES | O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10497890
|
Thermo Scientific Acros
124900010 |
1 kg | Plastic Bottle |
£45.10
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10743881
|
Thermo Scientific Acros
124900025 |
2.5 kg | Plastic Bottle |
£91.70
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10207113
|
Thermo Scientific Acros
124900100 |
10 kg | Plastic drum |
£247.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10034-99-8 | |
246.47 | |
WRUGWIBCXHJTDG-UHFFFAOYSA-L | |
24843 | |
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O |
H14MgO11S | |
MFCD00149785 | |
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate | |
CHEBI:31795 |
Specifications
770.0°C | |
White | |
Crystalline Powder and/or Crystals | |
H14MgO11S | |
MFCD00149785 | |
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate | |
WRUGWIBCXHJTDG-UHFFFAOYSA-L | |
magnesium;sulfate;heptahydrate | |
24843 | |
246.48 | |
Extra Pure | |
Magnesium sulfate heptahydrate |
10034-99-8 | |
5.0 to 9.2 (5% soln. at 20°C) | |
99% min. (on dry substance) (Complexometry) | |
MgSO4·7H2O | |
15, 5755 | |
Solubility in water: 710g/L (20°C). Other solubilities: soluble in glycerol,sparingly soluble in alcohol | |
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O | |
246.47 | |
CHEBI:31795 | |
≥99% | |
48.0% to 52.0% (450°C) |
RUO – Research Use Only