missing translation for 'onlineSavingsMsg'
Learn More

Magnesium sulfate heptahydrate, meets analytical spec. of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™

Catalog Number 15691250
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
6 x 2.5 kg
This item is not returnable. View return policy
10034-99-8
H14MgO11S
246.47
MFCD00149785
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
24843
CHEBI:31795
magnesium;sulfate;heptahydrate
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
10034-99-8
H14MgO11S
246.47
MFCD00149785
WRUGWIBCXHJTDG-UHFFFAOYSA-L
magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
24843
CHEBI:31795
magnesium;sulfate;heptahydrate
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 6 x 1 kg
Linear Formula MgSO4 · 7H2O
UN Number NONH for all modes of transport
Formula Weight 246.47
Percent Purity 99.5 to 100.5% (calc. to the dried substance)
Vapor Pressure 0.1mmHg (20°C)
Density Vapor Density: <0.01 (vs air)
Chemical Name or Material Magnesium Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'