missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, Heptahydrate, MP Biomedicals™

Catalog Number 11416581
Click to view available options
Quantity:
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Plays a variety of biological roles in enzymology, cell and nucleic acid structure, and muscle cell physiology. Magnesium Sulfate, Heptahydrate, MP Biomedicals is a common pharmaceutical preparation of magnesium, also known as Epsom salts. It is the main

Magnesium plays a role in cell membrane and cell wall structural integrity and in muscle cell physiology. It is an essential co-factor in many enzymes, including deoxyribonuclease (DNase), the restriction enzymes EcoR I and EcoR V, and Ribonuclease H. Magnesium also stabilizes polymeric nucleic acids, such as transfer RNA and ribozymes.

CAS 10034-99-8
Physical Form Crystalline Powder
Quantity 5 kg
Molecular Formula H14MgO11S
MDL Number MFCD00149785
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
Solubility Information Soluble in water (100mg/mL).
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
IUPAC Name magnesium;sulfate;heptahydrate
Molecular Weight (g/mol) 246.47
PubChem CID 24843
ChEBI CHEBI:31795
Formula Weight 246.47g/mol
Percent Purity ≥99%
Grade Molecular Biology
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'