missing translation for 'onlineSavingsMsg'
Learn More

Magnesium tert-butoxide, 93+%, Thermo Scientific Chemicals

Product Code. 10055553
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32149-57-8
Molecular Formula C8H18MgO2
Molecular Weight (g/mol) 170.53
MDL Number MFCD03788761
InChI Key RTKCPZYOLXPARI-UHFFFAOYSA-N
Synonym magnesium tert-butoxide, magnesium di-tert-butoxide, magnesium 2-methylpropan-2-olate, magnesium t-butoxide, magnesiumtert-butoxide, di-tert-butoxymagnesium, magnesium tert. butoxide, ksc226k7n, magnesium 2+ bis 2-methylpropan-2-olate
PubChem CID 12040442
IUPAC Name magnesium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].CC(C)(C)[O-].[Mg+2]

Specifications

Color White to Gray
Assay Percent Range 93+%
Packaging Glass Bottle
Quantity 10 g
Solubility Information Solubility in water: reacts violently
Formula Weight 170.53
Percent Purity 93+%
Physical Form Powder
Chemical Name or Material Magnesium tert-butoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'