missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium tungsten oxide, 99% (metals basis), Thermo Scientific™
£55.76 - £174.00
Chemical Identifiers
CAS | 13573-11-0 |
---|---|
Molecular Formula | MgO4W |
Molecular Weight (g/mol) | 272.141 |
MDL Number | MFCD00016209 |
InChI Key | DJZHPOJZOWHJPP-UHFFFAOYSA-N |
Synonym | magnesium tungstate, magnesium tungstate vi, magnesium tungsten oxide mgwo4, magnesium dioxido dioxo tungsten, magnesiumwolframat, magnesium wolframate, of a kind used as a luminophore, mg.wo4, tungstate wo42-, magnesium 1:1 , t-4, magnesium 2+ tungstate, magnesium tungstate-w |
PubChem CID | 3084161 |
IUPAC Name | magnesium;dioxido(dioxo)tungsten |
SMILES | [O-][W](=O)(=O)[O-].[Mg+2] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13573-11-0 | |
272.141 | |
DJZHPOJZOWHJPP-UHFFFAOYSA-N | |
3084161 | |
[O-][W](=O)(=O)[O-].[Mg+2] |
MgO4W | |
MFCD00016209 | |
magnesium tungstate, magnesium tungstate vi, magnesium tungsten oxide mgwo4, magnesium dioxido dioxo tungsten, magnesiumwolframat, magnesium wolframate, of a kind used as a luminophore, mg.wo4, tungstate wo42-, magnesium 1:1 , t-4, magnesium 2+ tungstate, magnesium tungstate-w | |
magnesium;dioxido(dioxo)tungsten |
Specifications
13573-11-0 | |
MgO4W | |
magnesium tungstate, magnesium tungstate vi, magnesium tungsten oxide mgwo4, magnesium dioxido dioxo tungsten, magnesiumwolframat, magnesium wolframate, of a kind used as a luminophore, mg.wo4, tungstate wo42-, magnesium 1:1 , t-4, magnesium 2+ tungstate, magnesium tungstate-w | |
[O-][W](=O)(=O)[O-].[Mg+2] | |
272.141 | |
272.16 | |
(metals basis) |
Powder | |
MFCD00016209 | |
DJZHPOJZOWHJPP-UHFFFAOYSA-N | |
magnesium;dioxido(dioxo)tungsten | |
3084161 | |
99% | |
Magnesium tungsten oxide |
Safety and Handling
missing translation for 'einecsNumber' : 236-999-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures