Learn More
Macherey-Nagel™ MagnetaPure 32 Plus Automated DNA / RNA Purification System
MagnetaPure 32 Plus automated DNA / RNA purification system: Automated extraction and purification system for DNA and RNA, MN.
Brand:  Macherey-Nagel™ 747010
Product Code. 18257428
  • £22910.00 / Each
Explore more special offers
Description
Description
  • The MagnetaPure 32 Plus is an automated platform for magnetic bead-based DNA/RNA extraction with a minimal benchtop footprint
  • Capable of processing up to 32 samples simultaneously
  • The stand-alone instrument is easy to set up, program, and operate
  • Mixing, magnetic bead transfer, washing, and elution steps are performed automatically, saving valuable hands-on time
  • The purification process can be completed within 20 to 60 minutes depending on the type of NucleoMag kit used and the complexity of the program
Specifications
Specifications
50 to 1000 μL
1 Piece
1 Piece
1 to 32 Samples
Automated extraction system for magnetic bead-based DNA/RNA purification