missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Magnetic Stirring Bar Set
Cylindrical stirring bars with smooth surface, many uses.Magnetic stirring bar set consists of 1 of the most standard bars, complete with crystal clear box
Brand: Buerkle™ 8271-1000
77.44 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Cylindrical stirring bars with smooth surface, many uses
- Magnetic stirring bar set consists of 10 of the most standard bars, complete with crystal clear box
- 1 of each 15 x 6 mm, 30 x 7mm
- 2 of each 20 x 7 mm, 25 x 7 mm, 40 x 7 mm, 50 x 7 mm
Specifications
White | |
Magnet | |
Cylinder |
Crystal clear box 1 of each 15 x 6 mm, 30 x 7 mm and 2 of each 20 x 7 mm, 25 x 7 mm, 40 x 7 mm, 50 x 7 mm | |
1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction