missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ MagniSort™ Mouse CD90.2 Positive Selection Kit
Designed for the magnetic separation of CD90.2+ cells by positive selection. It has been optimized for the isolation of CD90.2+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD90.2 antibody and streptavidin-coated magnetic beads.
Brand: Invitrogen™ 8802-6844-74
Alert:
Contains Sodium azide
Includes:
- MagniSort™ Anti-Mouse CD90.2 Biotin : 200 tests, 20μL/test
- MagniSort™ Positive Selection Beads A
Description
- CD90.2+ cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the undesired cells can be separated from CD90.2+ cells by decanting
- After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD90.2, clone 53-2.1
- When the starting population of CD90.2 cells is less than 5% of the total cells, the amount of MagniSort™ Magnetic Positive Selection Beads to be used can be reduced to 10 μL/test to maximize purity

Specifications
2° to 8°C. Do not freeze. | |
Magnetic Cell Separation | |
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
200 Tests |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ eBioscience™ MagniSort™ Mouse CD90.2 Positive Selection Kit > MagniSort Mouse CD90.2 Positive Selection Kit
Spot an opportunity for improvement?Share a Content Correction