Learn More
Invitrogen™ MagniSort™ Mouse Hematopoietic Lineage Depletion Kit
Designed for the magnetic depletion of mouse hematopoietic cells with committed lineages from mouse bone marrow. It uses a cocktail of antibody specific for hematopoietic lineages and streptavidin-coated magnetic beads.
Brand: Invitrogen™ 8804-6829-74
Alert:
Contains Sodium azide
Includes:
- MagniSort Mouse Hematopoietic Lineage Antibody Cocktail : 200 tests, 20μL/test
- MagniSort Negative Selection Beads B
Description
- Lineage committed cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the lineage negative cells can be separated from lineage committed cells by decanting
- After depletion, the enrichment of multi-potent progenitor cells can be verified by staining with Anti-Mouse CD34 (clone RAM34), and the enrichment of CD34- hematopoietic stem cell can be verified by additional staining with CD150 (clone mShad150), CD48 (clone HM48-1)
Anti-Mouse CD2 Biotin
Anti-Mouse CD3 Biotin
Anti-Mouse CD5 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD19 Biotin
Anti-Mouse CD45R Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse TER-119 Biotin
Specifications
Hematopoietic Cells | |
Cell Depletion Kit | |
200 tests | |
Mouse |
For Research Use Only. Not for use in diagnostic procedures.