missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAGPIX™ Calibration Kit

Product Code. 12383613
Click to view available options
Quantity:
25 Reactions, 25 rxn(s)
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 12383613

Brand: Invitrogen™ MPXCALK25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

This convenient calibration kit contains xMAP™ calibrators for the MAGPIX™ system classification and reporter channels.

This convenient calibration kit contains xMAP™ calibrators for the MAGPIX™ system classification and reporter channels. Calibration normalizes the settings for the system and helps ensure optimal and consistent microsphere classification. The kit is intended to be used with the off-plate reagent block. Each kit provides 25 calibrations.

Each Kit includes:

  • Stripwells
  • A lot-specific CD for importing target values

The MAGPIX™ system is a versatile, compact, fluorescent detection system based on Luminex's proven xMAP™ technology. The system is a simple immunoassay platform designed for the simultaneous analysis of up to 50 intracellular and extracellular markers in single biological samples. The MAGPIX™ system provides users with a powerful multiplexing tool that offers higher sample throughput, increased assay flexibility, reduced sample volume, and lower costs than traditional ELISA and Western blotting.

TRUSTED_SUSTAINABILITY

Specifications

Description MAGPIX™ Calibration Kit, 25 rxns, Calibration Standards Calibration Type, For Use With Luminex™ Instruments, MAGPIX™ System, Used For Protein Assays and Analysis, 2 to 8°C Storage Temperature, Wet Ice Shipping Condition
Product Line MAGPIX
Content And Storage • 25 strip wells (Cat. No. 13-52047)
• MAGPIX™ Calibrator Microspheres, 5 mL (MPXCAL-05)
• MAGPIX™ Calibration Kit CD (89-20233-00-001)

Store at 2°C to 8°C., 2 to 8°C
For Use With (Application) Protein assays and analysis
Includes 25 Strip wells (PN: 13-52047), MAGPIX™ calibrator microspheres, 5mL (PN: MPXCAL-05), MAGPIX™ calibration kit CD (PN: 89-20233-00-001)
Quantity 25 Reactions, 25 rxn(s)
Sample Volume 25 rxn(s)
Shipping Condition Wet Ice
Product Type MAGPIX™ Calibration Kit, Calibration Kit
For Use With (Equipment) Flow Cytometer

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.