missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ MAGPIX™ Drive Fluid (4 pack)
MAGPIX™ Drive Fluid (4 pack)
Brand: Novex™ MPXDF4PK
Code : 50
Additional Details : Weight : 3.00000kg
Description
MAGPIX™ Drive Fluid is the delivery medium for transporting the sample being analyzed on the Luminex™ MAGPIX™ system to the optic components. Each unit in the 4-pack of drive fluid contains 750 ml.Please refer to a Luminex™ instrument user manual for proper installation and use of MAGPIX™ Drive Fluid with the MAGPIX™ System.
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Specifications
Drive Fluid | |
MAGPIX™ | |
Room Temperature |
Luminex™ Instruments, MAGPIX™ System | |
4 x 750 mL |
For Research Use Only. Not for use in diagnostic procedures.