Learn More
Cytiva MagRack Maxi
A magnetic rack for small-scale protein purification and sample enrichment with magnetic beads
Brand: Cytiva 28-9864-41
Code : GH
Additional Details : Weight : 0.20000kg
Description
For use with Protein A Mag Sepharose, Protein A Mag Sepharose Xtra, Protein G Mag Sepharose, Protein G Mag Sepharose Xtra, His Mag Sepharose Ni, NHS Mag Sepharose, TiO2 Mag Sepharose, and Streptavidin Mag Sepharose.
- Anodized aluminum housing (blue) with a detachable plastic bar (white) containing a neodymium magnet
- Covers a broad range of sample volumes from low–microliter to high–milliliter purification scales
- Ability to vary the amount of medium as well as the sample volume
- Enrichment of sample volumes up to 50mL (1.7 oz.)
- Performs small-scale purifications
Specifications
Anodized Aluminum, Plastic |