missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences MagStrep 'Type3' XT Beads
MagStrep 'type3' XT beads are magnetic beads (5% suspension) to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Brand: IBA Lifesciences 2-4090-002
Code : NEW
Additional Details : Weight : 0.00080kg
Description
- MagStrep “type3” XT beads are magnetic beads consisting of a 6% agarose coated with the streptavidin variant Strep-Tactin™XT.
- Due to the magnetic core no centrifugation step is required since the beads can be easily and swiftly separated with a magnetic separator, reducing the risk of disturbing the pellet or loss of beads within the wash step to zero.
- As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein.
- MagStrep “type3” XT beads provide the highest binding capacity.
- These characteristics make them a perfect tool for isolation of proteins or complexes via pull-downs, immunoprecipitation (IP), or co-immunoprecipitation (Co-IP) in batch format and/or high-throughput screenings.
- The elution of target proteins or complexes can be triggered under mild conditions with the biotin-containing Buffer BXT or under denaturing conditions by boiling in SDS gel loading buffer.
- 20 μL suspension correspond to 1 μL magnetic beads.
Specifications
2°C to 8°C. Do not freeze. | |
Beads | |
3 |
Strep-Tactin Coated Magnetic Beads | |
2 mL |
For Research Use Only.