Learn More
BINDER Maintenance
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series B, BD, BD-S, E, ED, ED-S
BINDER Chambers Series BF, FD, FD-S, FED, FP, M, FDL
BINDER Chambers Series CB-S, CB
BINDER Chambers Series CBF
BINDER Chambers Series KBF, KMF
BINDER Chambers Series KBF-S, KBF-S ECO
BINDER Chambers Series KT, KB, KB ECO
BINDER Chambers Series MK, MKT
BINDER Chambers Series MKF, MKFT
BINDER Chambers Series UF V
BINDER Chambers Series VD
BINDER Chambers Series VDL
Unit Size:
Each
Description
One-off maintenance service in accordance with maintenance schedule.
- Visual inspection of mechanical and electrical components, testing of all key functions.
- Calibration of a test temperature in the center of usable space as specified by the user
- Without certificate
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers Series MK, MKT |
| Product Type | Preventive Maintenance |