missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ S-Series Hood with Faceseal, Integrated Head Suspension
Versaflo™ S-Series Hood with integrated suspension provides head, face, neck, and shoulder coverage as well as providing eye and face protection from liquid splash and low energy flying particles and dust.
£115.00 - £419.00
Specifications
Certifications/Compliance | NIOSH |
---|---|
Color | White |
For Use With (Application) | Assembly, facility maintenance, machine operation, painting, sanding |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
10357839
|
3M™
7000044302 |
S-433 S |
£115.00
Each |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
10450180
|
3M™
7000000145 |
S-433 L |
£115.00
Each |
In Stock
Log in to see stock. |
|||||
11771153
|
3M™
7100011287 |
S-533 S |
£138.00
Each |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
11761153
|
3M™
7000000147 |
S-533 L |
£136.00
Each |
In Stock
Log in to see stock. |
|||||
11810114
|
3M™
7100015401 |
S-433 L-5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Integrated head suspension provides head, face, neck and shoulder coverage.
- Protects face and eyes from liquid splash and low energy flying particles and dust (Grade F).
- Suitable for applications which require frequent replacement of the entire headtop assembly or those where there is less soiling of the outer fabric.
- No assembly required
- Ready to use straight out of the box.
Specifications
NIOSH | |
White | |
Assembly, facility maintenance, machine operation, painting, sanding |