missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Maintenance Free Half Mask
This Maintenance Free Half Mask is a simple and ready-to-use respirator with an integrated cartridge and filters.
Brand: 3M™ 7100113101
22.95 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Additional Details : Weight : 0.27900kg
Description
- Effective and comfortable protection against organic vapor, inorganic vapor, acid gases, ammonia, and hazardous particulates.
- A newly enhanced valve assembly helps to reduce exhalation breathing resistance for improved breathing comfort.
- Lightweight, well-balanced, low profile design.
- Integrated cartridge and filters mean it's ready to use and maintenance free.
- Soft, close-fitting face seal made with textured, non-allergenic material.
- Includes a resealable bag for hygienic storage and transportation.
Specifications
FFA2P3 R D | |
EN 405:2001+A1:2009 | |
Blue | |
Reusable | |
Thermoplastic Elastomer | |
One Size | |
TPE | |
FFA2P3 R D Filters | |
4255+ |
EN 405:2001, A1:2009 | |
Permanent | |
Half Mask | |
Bonded Carbon | |
Reusable Respirator | |
Yes | |
Polyphenylene Ether | |
Polyisoprene |