Learn More
3M™ Maintenance-Free Reusable Half Mask 4000+ Series
Available to help prolong the life of the main particulate filter for the 3M™ Maintenance-Free Reusable Mask 4000+ Series. Traps larger spray particles before they reach and clog the filters.
Brand:  3M™ 7100113104
483.89 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 17257650
  • £977.00 / Pack of 500
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 3M™ Overspray Guard 400+ prolongs respirator service life by reducing clogging of the particulate filters during paint spray applications
  • Keeps filters clean during paint spray applications
  • Traps larger spray particles before they reach and clog the filters
  • For use with 3M™ Reusable Respirator 4000+ Series
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Reusable
3M™ Maintenance-Free Reusable Mask 4000+ Series