missing translation for 'onlineSavingsMsg'
Learn More

3M™ Maintenance Sorbent Mini-Boom

Can be placed around the base of equipment to help contain fluids that may leak or spill, preventing spread to adjacent work areas.

£187.00 - £194.00

Specifications

Color Gray
Capacity (English) 11.89 gal.
Type Sorbent Mini Boom
View More Specs

Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
17138600
View Documents
3M™
7000051876
240 cm
£187.00
Pack of 6
Estimated Shipment: 06-01-2025
to see stock.
Add to basket
 
17128600
View Documents Promotion Details
3M™
7000051875
370 cm
£194.00
Pack of 4
Estimated Shipment: 08-01-2025
to see stock.
Add to basket
 
Description

Description

  • Place around equipment to contain spills and prevent leaks from spreading to adjacent work areas
  • For use on most non-aggressive liquids
  • Promotes a cleaner, safer working environment
  • Very easy to use and to retrieve
Specifications

Specifications

Gray
11.89 gal.
Sorbent Mini Boom
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.