missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Malate Dehydrogenase Activity Assay Kit (Fluorometric)

This kit provides a quick and easy method for monitoring MDH activities in a wide variety of samples.

Brand:  Novus Biologicals™ NBP2-59750

Product Code. 18202021

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

In this assay, MDH converts malate into intermediate products, which will further react with the Probe forming a fluorophore. Detection method: Fluorescence (Ex/Em = 535/587nm). The kit is simple, sensitive and high-throughput adaptable and can detect as low as 40 uU of MDH activity.

  • Suitable for the analysis of citric acid cycle and malate-asparate shuttle
  • Can be used with animal heart, liver, muscle tissues
  • Can be used with purified mitochondria
  • Can be used with adherent or suspension cell cultures

Kit Contents:

  • Probe
  • NADH Standard
  • MDH Assay Buffer
  • MDH Developer
  • MDH Positive Control
  • MDH Substrate
Specifications

Specifications

100 Assays
Assay Kit (Fluorometric)
Store at -20°C in the dark. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.