Learn More
Thermo Scientific™ Male Luer-LOK Priming Syringes
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
Brand:  Thermo Scientific™ 365NL561
 View more versions of this product
Product Code. 10585995
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gas Tight GC Autosampler Syringe, Luer-Lok
10 mL
Gas Chromatography Systems
365RNL22
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.