Learn More
Idex Male-Threaded Nuts
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
Brand:  Idex U-400X
Encompass
 View more versions of this product
Product Code. 13070950
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
  • Eliminates tubing twist that can loosen connections
  • Resistant to loosening through vibration
Specifications
Specifications
Nut
0.4 in.
1/16 in. O.D. Tubing
0.0625 in.
0.25 in.
Stainless Steel