Learn More
Hamilton™ Male Tube, Stainless Steel Tube Adapter 1/4-28, 1/16in.
Product Code. 10515673
Click to view available options
Packaging Quantity:
1 Each
Unit Size:
Each
Product Code. 10515673
Brand: Hamilton™ 32805
Only null left
Add to basket
Add to basket
Suitable adapter for 1/4-28 threaded valve ports, OEM liquid handling tubing.
  • Male tube
  • Stainless steel tube adapter
  • 1/4-28 threaded valve ports, OEM liquid handling tubing
Specifications
Type Male Tube, Stainless Steel Tube Adapter
Diameter (English) Outer 1/16 in.
Packaging Quantity 1 Each
For Use With 1/4-28 threaded valve ports, OEM liquid handling tubing
Material Stainless Steel