missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maleate, 0.2M buffer soln., pH 5.5, Thermo Scientific™
£44.10 - £67.80
Chemical Identifiers
CAS | 3105-55-3 |
---|---|
Molecular Formula | C4H2NaO4 |
Molecular Weight (g/mol) | 137.05 |
MDL Number | MFCD00070246 |
InChI Key | VRVKOZSIJXBAJG-ODZAUARKSA-L |
PubChem CID | 91658932 |
SMILES | [Na+].[O-]C(=O)\C=C/C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15424509
|
Thermo Scientific Alfa Aesar
J62049.AK |
250 mL |
£44.10
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15434509
|
Thermo Scientific Alfa Aesar
J62049.AP |
500 mL |
£67.80
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
3105-55-3 | |
137.05 | |
VRVKOZSIJXBAJG-ODZAUARKSA-L | |
[Na+].[O-]C(=O)\C=C/C([O-])=O |
C4H2NaO4 | |
MFCD00070246 | |
91658932 |
Specifications
3105-55-3 | |
0.2 M | |
Liquid | |
MFCD00070246 | |
[Na+].[O-]C(=O)\C=C/C([O-])=O | |
137.05 | |
Maleate buffer soln. |
Colorless | |
5.5 | |
C4H2NaO4 | |
VRVKOZSIJXBAJG-ODZAUARKSA-L | |
sodium;(Z)-4-hydroxy-4-oxobut-2-enoate;trihydrate | |
91658932 |
Safety and Handling
missing translation for 'einecsNumber' : 221-461-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures