missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maleate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£46.55 - £96.60
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12952145
|
Fisher Chemical
A 026 100 |
100 mL | Glass Bottle |
£46.55
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12962145
|
Fisher Chemical
A 026 250 |
250 mL | Glass Bottle |
£66.35
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12972145
|
Fisher Chemical
A 026 500 |
500 mL | Glass Bottle |
£96.60
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
C2H2(COO)22- |
Specifications
1g/L | |
C2H2(COO)22- | |
Liquid |
Single-component Standard Solution | |
H2O | |
Maleate |
Safety and Handling
H317
P261;P272;P280;P302+P352;P333+P313;P362+P364;P501
Warning
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-component Standard Solution for Ion chromatography. Maleate - C2H2(COO)22- 1000mg/l in water (A 026)
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .