Learn More
Maleic Acid, 99%, Thermo Scientific Chemicals
£26.33 - £888.42
Chemical Identifiers
CAS | 110-16-7 |
---|---|
Molecular Formula | C4H4O4 |
Molecular Weight (g/mol) | 116.07 |
MDL Number | MFCD00063177 |
InChI Key | VZCYOOQTPOCHFL-OWOJBTEDSA-N |
Synonym | maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid |
PubChem CID | 444266 |
ChEBI | CHEBI:18300 |
IUPAC Name | (Z)-but-2-enedioic acid |
SMILES | OC(=O)\C=C\C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10396760
|
Thermo Scientific
125231000 |
100 g | Plastic bottle |
£26.33
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10439050
|
Thermo Scientific Acros
125230010 |
1 kg | Plastic bottle |
£44.22
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10348843
|
Thermo Scientific
125230051 |
5 kg | Plastic drum |
£171.86
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10033853
|
Thermo Scientific Acros
125230250 |
25 kg | Fibre drum |
£888.42
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-16-7 | |
116.07 | |
VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
444266 | |
(Z)-but-2-enedioic acid |
C4H4O4 | |
MFCD00063177 | |
maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid | |
CHEBI:18300 | |
OC(=O)\C=C\C(O)=O |
Specifications
110-16-7 | |
White | |
C4H4O4 | |
MFCD00063177 | |
15, 5767 | |
Solubility in water: 790g/L (25°C). Other solubilities: soluble in glacial acetic acid, soluble in alcohol and acetone, slightly soluble in ether and benzene, practically insoluble in carbon tetrachloride | |
OC(=O)\C=C\C(O)=O | |
116.07 | |
CHEBI:18300 | |
99% | |
Authentic | |
Maleic acid |
134.0°C to 138.0°C | |
99% | |
HO2CCH=CHCO2H | |
02, IV, 2199 | |
maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid | |
VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
(Z)-but-2-enedioic acid | |
444266 | |
116.07 | |
127°C | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
- Maleic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.