Learn More
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
£18.29 - £321.60
Chemical Identifiers
CAS | 108-31-6 |
---|---|
Molecular Formula | C4H2O3 |
Molecular Weight (g/mol) | 98.06 |
MDL Number | MFCD00005518 |
InChI Key | FPYJFEHAWHCUMM-UHFFFAOYSA-N |
Synonym | maleic anhydride, 2,5-furandione, maleic acid anhydride, toxilic anhydride, dihydro-2,5-dioxofuran, cis-butenedioic anhydride, 2,5-dihydrofuran-2,5-dione, polymaleic anhydride, maleinanhydrid, rcra waste number u147 |
PubChem CID | 7923 |
ChEBI | CHEBI:474859 |
SMILES | O=C1OC(=O)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12694317
|
Thermo Scientific Acros
125240250 |
25 g | Plastic bottle |
£18.29
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12634937
|
Thermo Scientific Acros
125240010 |
1 kg | Plastic bottle |
£22.71
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12695995
|
Thermo Scientific Acros
125240050 |
5 kg | Plastic drum |
£72.66
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12654937
|
Thermo Scientific Acros
125240251 |
25 kg | Fibre drum |
£321.60
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-31-6 | |
98.06 | |
FPYJFEHAWHCUMM-UHFFFAOYSA-N | |
7923 | |
O=C1OC(=O)C=C1 |
C4H2O3 | |
MFCD00005518 | |
maleic anhydride, 2,5-furandione, maleic acid anhydride, toxilic anhydride, dihydro-2,5-dioxofuran, cis-butenedioic anhydride, 2,5-dihydrofuran-2,5-dione, polymaleic anhydride, maleinanhydrid, rcra waste number u147 | |
CHEBI:474859 |
Specifications
108-31-6 | |
White | |
200.0°C | |
Authentic | |
C4H2O3 | |
17, V,11, 55 | |
15,5768 | |
maleic anhydride, 2,5-furandione, maleic acid anhydride, toxilic anhydride, dihydro-2,5-dioxofuran, cis-butenedioic anhydride, 2,5-dihydrofuran-2,5-dione, polymaleic anhydride, maleinanhydrid, rcra waste number u147 | |
FPYJFEHAWHCUMM-UHFFFAOYSA-N | |
furan-2,5-dione | |
7923 | |
1.6 mPa.s (60°C) | |
99% | |
Pellets |
52.0°C to 55.0°C | |
1.4800g/mL | |
102°C | |
99% | |
MFCD00005518 | |
04,316; 05,422 | |
1.48 | |
Solubility in water: 790g/L (25°C). Other solubilities: reacts with water forming maleic acid,227g/100g acetone- 50g/100g benzene (25°C),112g/100g ethylacetate (25°C)- 0,60g/100g ccl4,52.5g/100g chloroform (25°C)-23,4g/100g toluen | |
O=C1OC(=O)C=C1 | |
98.06 | |
CHEBI:474859 | |
98.06 | |
Reagent | |
Maleic anhydride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes damage to organs through
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-571-6
- Maleic anhydride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.