missing translation for 'onlineSavingsMsg'
Learn More

Malonaldehyde bis(phenylimine) monohydrochloride, 97+%

Product Code. 11470327
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11470327

Brand: Thermo Scientific Alfa Aesar B21721.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123071-42-1 | C15H15ClN2 | 258.749 g/mol

Malonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonaldehyde bis(phenylimine) monohydrochloride is a useful fluorescent cyanine dye.

Solubility
Soluble in water (partly).

Notes
Hygroscopic, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123071-42-1
Molecular Formula C15H15ClN2
Molecular Weight (g/mol) 258.749
MDL Number MFCD00054634
InChI Key FLMLOZWXXPLHIE-UHFFFAOYSA-N
Synonym malonaldehyde bis phenylimine monohydrochloride, malonaldehyde bis phenylimine hydrochloride, malondialdehyde bis phenylimine monohydrochloride, malonaldehyde bis phenylimine hcl, malonaldehydedianilidehydrochloride, n-3-phenylimino-1-propen-1-yl aniline hydrochloride, n1,n3-diphenylpropane-1,3-diimine hydrochloride, 1e,3e-n1,n3-diphenylpropane-1,3-diimine hydrochloride
PubChem CID 12213307
IUPAC Name N,N'-diphenylpropane-1,3-diimine;hydrochloride
SMILES C1=CC=C(C=C1)N=CCC=NC2=CC=CC=C2.Cl

Specifications

Melting Point ∽215°C (decomposition)
Quantity 100 g
Solubility Information Soluble in water (partly).
Formula Weight 258.75
Percent Purity 98%
Chemical Name or Material Malonaldehyde bis(phenylimine) monohydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.