Learn More
Malonic acid, 99%, Thermo Scientific Chemicals
£71.70 - £742.00
Chemical Identifiers
CAS | 141-82-2 |
---|---|
Molecular Formula | C3H4O4 |
Molecular Weight (g/mol) | 104.06 |
InChI Key | OFOBLEOULBTSOW-UHFFFAOYSA-N |
Synonym | malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid |
PubChem CID | 867 |
ChEBI | CHEBI:30794 |
IUPAC Name | propanedioic acid |
SMILES | C(C(=O)O)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10113320
|
Thermo Scientific Acros
125262500 |
250 g | Plastic bottle |
£71.70
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10316770
|
Thermo Scientific
125260010 |
1 kg | Plastic bottle |
£205.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11955021
|
Thermo Scientific Acros
125260051 |
5 kg | Plastic bottle |
£742.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-82-2 | |
104.06 | |
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid | |
CHEBI:30794 | |
C(C(=O)O)C(=O)O |
C3H4O4 | |
OFOBLEOULBTSOW-UHFFFAOYSA-N | |
867 | |
propanedioic acid |
Specifications
141-82-2 | |
White | |
Authentic | |
C3H4O4 | |
02,566 | |
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid | |
OFOBLEOULBTSOW-UHFFFAOYSA-N | |
propanedioic acid | |
867 | |
104.06 | |
Crystalline Powder |
130.0°C to 135.0°C | |
172°C | |
99% | |
HO2CCH2CO2H | |
15,5774 | |
Solubility in water: 1400g/L (20°C). Other solubilities: soluble in alcohol and ether | |
C(C(=O)O)C(=O)O | |
104.06 | |
CHEBI:30794 | |
99% | |
Malonic acid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-503-
- Malonic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only