missing translation for 'onlineSavingsMsg'
Learn More

Malonic acid disodium salt, 99%, Thermo Scientific Chemicals

Product Code. 11356249
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Malonic acid disodium salt is used in organic Synthesis, pharmaceuticals, agrochemicals and dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-95-7
Molecular Formula C3H2Na2O4
Molecular Weight (g/mol) 148.025
MDL Number MFCD00002708
InChI Key PRWXGRGLHYDWPS-UHFFFAOYSA-L
Synonym sodium malonate, disodium malonate, propanedioic acid, disodium salt, malonic acid disodium salt, disodium propanedioate, malonic acid, sodium salt, unii-9qwe64m39h, propanedioic acid, sodium salt 1:2, malonic acid, disodium salt, dicarboxymethane disodium salt
PubChem CID 8865
ChEBI CHEBI:62983
IUPAC Name disodium;propanedioate
SMILES C(C(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Quantity 100 g
Solubility Information Soluble in water (148g/L at 20°C).
Formula Weight 148.03
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Malonic acid disodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.