missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malonic acid disodium salt hydrate, 99%, Thermo Scientific Chemicals
£28.50 - £232.00
Chemical Identifiers
CAS | 132499-82-2 |
---|---|
Molecular Formula | C3H2Na2O4 |
Molecular Weight (g/mol) | 148.03 |
MDL Number | MFCD00149114 |
InChI Key | PRWXGRGLHYDWPS-UHFFFAOYSA-L |
IUPAC Name | disodium propanedioate |
SMILES | [Na+].[Na+].[O-]C(=O)CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10645501
|
Thermo Scientific Acros
413500250 |
25 g | Plastic bottle |
£28.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10685681
|
Thermo Scientific Acros
413501000 |
100 g | Plastic bottle |
£63.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10165503
|
Thermo Scientific Acros
413505000 |
500 g | Plastic bottle |
£232.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
132499-82-2 | |
148.03 | |
PRWXGRGLHYDWPS-UHFFFAOYSA-L | |
[Na+].[Na+].[O-]C(=O)CC([O-])=O |
C3H2Na2O4 | |
MFCD00149114 | |
disodium propanedioate |
Specifications
132499-82-2 | |
Authentic | |
C3H2Na2O4 | |
MFCD00149114 | |
PRWXGRGLHYDWPS-UHFFFAOYSA-L | |
disodium propanedioate | |
148.03 | |
Crystalline Powder |
White | |
99% | |
CH2(COONa)2·xH2O | |
Solubility in water: soluble | |
[Na+].[Na+].[O-]C(=O)CC([O-])=O | |
148.03 | |
99% | |
Malonic acid disodium salt hydrate |
RUO – Research Use Only