missing translation for 'onlineSavingsMsg'
Learn More

Melford Maltose

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Brand:  Melford M22000-250.0

 View more versions of this product

Product Code. 30018462

  • £51.45 / 250g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

CAS Number: 6363-53-7

Molecular Formula: C12H22O11H2O

Molecular Weight: 360.3
Specifications

Specifications

6363-53-7
C12H22O11-H2O
250 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.