missing translation for 'onlineSavingsMsg'
Learn More

Melford Maltose

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Brand:  Melford M22000-500.0

 View more versions of this product

Product Code. 30018463

  • £93.15 / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

CAS Number: 6363-53-7

Molecular Formula: C12H22O11H2O

Molecular Weight: 360.3
Specifications

Specifications

6363-53-7
C12H22O11-H2O
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers