missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Maltotriose, 98%
Click to view available options
Quantity:
1 g
5 g
Description
Maltotriose is a matrose regulon inducer in Escherichia coli. Maltotriose has been used in a study to investigate the influence of fourth generation poly(propyleneimine) dendrimers on blood cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Maltotriose is a matrose regulon inducer in Escherichia coli. Maltotriose has been used in a study to investigate the influence of fourth generation poly(propyleneimine) dendrimers on blood cells.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Maltotriose is a matrose regulon inducer in Escherichia coli. Maltotriose has been used in a study to investigate the influence of fourth generation poly(propyleneimine) dendrimers on blood cells.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 132°C to 135°C |
Color | White |
Quantity | 1 g |
Beilstein | 1443481 |
Solubility Information | Soluble in water. |
Optical Rotation | +165° (c=1 in Water) |
Formula Weight | 504.44 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Maltotriose |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'