Learn More
IBA Lifesciences pCSG-IBA Mammalian Expression Vector with Secretion
GREENER_CHOICE
Designed for high-level episomal expression in mammalian hosts under G418 selection. Mammalian Expression Vector with Secretion is a large expression vector with universal features for transient expression as well as for generation of stable cell lines (NeoR).
Brand:  IBA Lifesciences 5-5142-001
 View more versions of this product
Product Code. 15683868
  • £232.00 / 5µg
Explore more special offers
Description
Description
pCSG-IBA mammalian expression vectors
  • Designed for high-level episomal expression in mammalian hosts under G418 selection containing the following elements:
  • Human cytomegalovirus (CMV) immediate-early promoter for high-level expression in a wide range of mammalian cells
  • Episomal replication through Epstein Barr Virus replication origin (oriP) and nuclear antigen encoded by EBNA-1 in human, primate and canine cells and through SV40 replication origin in cells latently infected with SV40 or that express the SV40 large T antigen (e.g. HEK293T, COS-1, COS-7)
  • Stable cell lines can be selected by the neomycin resistance gene (NeoR).
  • In addition, the human cytomegalovirus (CMV) immediate-early promoter enables a high-level expression in a wide range of mammalian cells.
Please note that cloning into pCSG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. Besides to the direct cloning of the gene of interest into pCSG-IBA vectors with Esp3I, another option via a so-called entry-vector is possible.
Specifications
Specifications
Constitutive
CMV
Geneticin™ (G-418)
His Tag (6x), Twin-Strep-tag
pCSG-IBA142
Ampicillin
Vector
Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage
5 μg
Episomal expression (oriP, EBNA-1); BM40 secretion signal