Learn More
Emma Safety Footwear Mammoet M1807 XD Safety Shoes
Extra high boot is heat and cold insulated. Ankle protection Comfort Sponge with added padding provides a more comfortable feel and better fit. Mammoet M1807 is very easy to put on and take off; the vertical eyelets on the shoe make lacing the fire-retardant laces very simple.
Brand:  Emma Safety Footwear 765868-40
 View more versions of this product
Product Code. 15929031
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • GripForce™ Duo Heat Resistant Outsole
  • Heat and cold insulated
  • Fire-retardant laces
  • Waterproof Oil Nubuck construction
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
EN ISO 20345:2011
Black
Hydro-Tec™ Sanitized Silver / Sympatex™
40
Laces
Mammoet M1807 XD
Oiled Nubuck
Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Emma Safety Footwear Mammoet M1807 XD Safety Shoes > Size: 40