missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Black-Brown to Brown-Gray |
Assay Percent Range | 99+% |
Linear Formula | Mn |
Quantity | 1 kg |
Merck Index | 15, 5788 |
Formula Weight | 54.94 |
Percent Purity | ≥99.0% (Mn) |
Packaging | Glass bottle |
Form | Powder |
Chemical Name or Material | Manganese, 99+% |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction