missing translation for 'onlineSavingsMsg'
Learn More

Manganese acetate tetrahydrate, 99.999%, (trace metal basis)

Product Code. 10577200 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
Packaging:
Plastic bottle
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 10577200

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6156-78-1 | C4H14MnO8 | 245.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

Melting Point >300.0°C
CAS 6156-78-1
Color Pink
Physical Form Crystalline Powder and/or Crystals
Quantity 50 g
Assay Percent Range 21.8 to 23% (Complexometry)
Molecular Formula C4H14MnO8
Linear Formula (CH3CO2)2Mn·4H2O
MDL Number MFCD00062552
Flash Point >130°C
Merck Index 14,5724
Synonym manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in alcohol
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
IUPAC Name manganese(2+);diacetate;tetrahydrate
Molecular Weight (g/mol) 245.09
PubChem CID 93021
Formula Weight 245.09
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Manganese acetate tetrahydrate
Show More Show Less
Product Identifier
  • Manganese(II) acetate tetrahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.