missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) 2,4-pentanedionate, Thermo Scientific Chemicals

Product Code. 11443038
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14024-58-9
Molecular Formula C10H14MnO4
Molecular Weight (g/mol) 253.16
MDL Number MFCD00000022 MFCD09998212
InChI Key ZQZQURFYFJBOCE-FDGPNNRMSA-L
Synonym bis 4-hydroxypent-3-en-2-one dihydrate manganese
PubChem CID 54669727
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;manganese;dihydrate
SMILES [Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 260°C to 262°C (decomposition)
Density 1.6
Quantity 100 g
Formula Weight 253.16
Chemical Name or Material Manganese(II) 2,4-pentanedionate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'