Learn More
Manganese(II) 2,4-pentanedionate, Thermo Scientific Chemicals
£22.30 - £188.00
Chemical Identifiers
CAS | 14024-58-9 |
---|---|
Molecular Formula | C10H14MnO4 |
Molecular Weight (g/mol) | 253.16 |
MDL Number | MFCD00000022 MFCD09998212 |
InChI Key | ZQZQURFYFJBOCE-FDGPNNRMSA-L |
Synonym | bis 4-hydroxypent-3-en-2-one dihydrate manganese |
PubChem CID | 54669727 |
IUPAC Name | (Z)-4-hydroxypent-3-en-2-one;manganese;dihydrate |
SMILES | [Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11433038
|
Thermo Scientific Alfa Aesar
A18762.14 |
25 g |
£22.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443038
|
Thermo Scientific Alfa Aesar
A18762.22 |
100 g |
£53.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453038
|
Thermo Scientific Alfa Aesar
A18762.36 |
500 g |
£188.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14024-58-9 | |
253.16 | |
ZQZQURFYFJBOCE-FDGPNNRMSA-L | |
54669727 | |
[Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
C10H14MnO4 | |
MFCD00000022 MFCD09998212 | |
bis 4-hydroxypent-3-en-2-one dihydrate manganese | |
(Z)-4-hydroxypent-3-en-2-one;manganese;dihydrate |
Specifications
14024-58-9 | |
1.6 | |
MFCD00000022 MFCD09998212 | |
ZQZQURFYFJBOCE-FDGPNNRMSA-L | |
(Z)-4-hydroxypent-3-en-2-one;manganese;dihydrate | |
54669727 | |
Manganese(II) 2,4-pentanedionate |
260°C to 262°C (decomposition) | |
C10H14MnO4 | |
bis 4-hydroxypent-3-en-2-one dihydrate manganese | |
[Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
253.16 | |
253.16 |
Safety and Handling
GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H351
missing translation for 'einecsNumber' : 237-858-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only