missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) acetate, anhydrous, 98+%

Product Code. 11460128
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11460128

Brand: Thermo Scientific Alfa Aesar B23708.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 638-38-0 | C4H6MnO4 | 173.03 g/mol

Manganese(II) acetate is used as a dye mordant and varnish drier. It is also used in fertilizers, food packaging and feed additives. Further, it serves as a desiccant, catalyst, intermediate and ion exchange agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) acetate is used as a dye mordant and varnish drier. It is also used in fertilizers, food packaging and feed additives. Further, it serves as a desiccant, catalyst, intermediate and ion exchange agent.

Solubility
Soluble in water, methanol and acetic acid.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong bases and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-38-0
Molecular Formula C4H6MnO4
Molecular Weight (g/mol) 173.03
MDL Number MFCD00013039
InChI Key UOGMEBQRZBEZQT-UHFFFAOYSA-L
Synonym manganese ii acetate, manganese acetate, manganous acetate, diacetylmanganese, manganese diacetate, manganese di acetate, manganese 2+ acetate, octan manganaty czech, acetic acid, manganese 2+ salt, unii-0v6e9q2i0y
PubChem CID 12525
IUPAC Name manganese(2+);diacetate
SMILES [Mn++].CC([O-])=O.CC([O-])=O

Specifications

Linear Formula Mn(OOCCH3)2
Quantity 1000 g
Sensitivity Hygroscopic
Merck Index 14,5724
Solubility Information Soluble in water,methanol and acetic acid.
Formula Weight 173.03
Percent Purity ≥98%
Chemical Name or Material Manganese(II) acetate, anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.