missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) Acetate Tetrahydrate, ≥99.0% (KT), Honeywell™

Product Code. 15646090
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a.,≥99.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6156-78-1
Molecular Formula C4H14MnO8
Molecular Weight (g/mol) 245.09
MDL Number MFCD00062552
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
Synonym manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
PubChem CID 93021
IUPAC Name manganese(2+);diacetate;tetrahydrate
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 80°C
Color Pink
pH Acidic
Physical Form Solid
Quantity 250 g
Linear Formula (CH3COO)2Mn · 4H2O
UN Number NONH for all modes of transport
Beilstein 3730728
Formula Weight 245.09g/mol
Percent Purity ≥99.0% (KT)
Odor Vinegar-like
Packaging Poly Bottle
Density 1.58g/cm3
Chemical Name or Material Manganese(II) acetate tetrahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'