missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) Acetate Tetrahydrate, ≥99.0% (KT), Honeywell™
Purum p.a.,≥99.0%
£20.09 - £67.01
Chemical Identifiers
CAS | 6156-78-1 |
---|---|
Molecular Formula | C4H14MnO8 |
Molecular Weight (g/mol) | 245.09 |
MDL Number | MFCD00062552 |
InChI Key | CESXSDZNZGSWSP-UHFFFAOYSA-L |
Synonym | manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp |
PubChem CID | 93021 |
IUPAC Name | manganese(2+);diacetate;tetrahydrate |
SMILES | O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
6156-78-1 | |
245.09 | |
CESXSDZNZGSWSP-UHFFFAOYSA-L | |
93021 | |
O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O |
C4H14MnO8 | |
MFCD00062552 | |
manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp | |
manganese(2+);diacetate;tetrahydrate |
Specifications
80°C | |
Pink | |
Solid | |
(CH3COO)2Mn · 4H2O | |
NONH for all modes of transport | |
manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp | |
O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O | |
245.09 | |
245.09g/mol | |
Vinegar-like | |
Manganese(II) acetate tetrahydrate |
6156-78-1 | |
Acidic | |
C4H14MnO8 | |
MFCD00062552 | |
3730728 | |
CESXSDZNZGSWSP-UHFFFAOYSA-L | |
manganese(2+);diacetate;tetrahydrate | |
93021 | |
≥99.0% (KT) | |
1.58g/cm3 |
Safety and Handling
P261-P305 + P351 + P338
missing translation for 'rtecsNumber' : 211-334-3