Learn More
Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
£49.25 - £125.63
Chemical Identifiers
CAS | 6156-78-1 |
---|---|
Molecular Formula | C4H14MnO8 |
Molecular Weight (g/mol) | 245.09 |
MDL Number | MFCD00062552 |
InChI Key | CESXSDZNZGSWSP-UHFFFAOYSA-L |
Synonym | manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp |
PubChem CID | 93021 |
IUPAC Name | manganese(2+);diacetate;tetrahydrate |
SMILES | O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386907
|
Thermo Scientific Alfa Aesar
010802.06 |
5 g |
£49.25
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396907
|
Thermo Scientific Alfa Aesar
010802.14 |
25 g |
£125.63
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used as a desiccant, fertilizer and a catalyst. Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst. Used for the production of high purity manganese dioxide, manganese oxide hydrate, oxide-based catalysts, and catalysts for oxidation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6156-78-1 | |
245.09 | |
CESXSDZNZGSWSP-UHFFFAOYSA-L | |
93021 | |
O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O |
C4H14MnO8 | |
MFCD00062552 | |
manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp | |
manganese(2+);diacetate;tetrahydrate |
Specifications
80°C | |
Pink | |
C4H14MnO8 | |
MFCD00062552 | |
manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp | |
CESXSDZNZGSWSP-UHFFFAOYSA-L | |
manganese(2+);diacetate;tetrahydrate | |
93021 | |
99.999% | |
Pungent | |
(metals basis) |
6156-78-1 | |
Crystalline Aggregates | |
Mn(OOCCH3)2·4H2O | |
14,5724 | |
Soluble in water,methanol,ethanol,and acetic acid. | |
O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O | |
245.09 | |
245.09 (173.03 Anhydrous) | |
Puratronic™ | |
1.589 g/mL | |
Manganese(II) acetate tetrahydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c
H315-H319-H335-H336-H373
missing translation for 'einecsNumber' : 211-334-3
missing translation for 'rtecsNumber' : AI5775000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only