missing translation for 'onlineSavingsMsg'
Learn More
Manganese(II) bromide hydrate, 98%, Thermo Scientific Chemicals Product Code.: 11383758

Manganese(II) bromide hydrate, 98%, Thermo Scientific Chemicals

Product Code. 11383758
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11383758

Brand: Thermo Scientific Alfa Aesar 040245.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis. It can be used in place of palladium in the stille reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-20-6
Molecular Formula Br2H8MnO4
Molecular Weight (g/mol) 286.81
MDL Number MFCD00149791
InChI Key HHDPJRSXPOGIOP-UHFFFAOYSA-L
Synonym manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide
IUPAC Name manganese(2+) tetrahydrate dibromide
SMILES O.O.O.O.[Mn++].[Br-].[Br-]

Specifications

Melting Point 64.3°C (decomposition)
Color Pink
Physical Form Crystalline Aggregates
Quantity 25 g
Merck Index 14,5725
Solubility Information Soluble in water, alcohol
Formula Weight 214.76 (Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Manganese(II) bromide hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(II) bromide hydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.